ADVH1F1C5BDF AChemBlock

ethyl 1,4-dioxaspiro[4.5]decane-8-carboxylate

  • MDL number MFCD00205582
  • Cas Number : 1489-97-0
  • Molecular Weight : 214.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-1-4-dioxaspiro-4-5-decane-8-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P48012
Availability details for
Quantity
  • Availability
  • Add to Cart