ADVH1F1C5C0F AChemBlock

(1S,4S)-2,5-diazabicyclo[2.2.1]heptane dihydrochloride

  • MDL number MFCD28098133
  • Cas Number : 5260-20-8
  • Molecular Weight : 171.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1s-4s-2-5-diazabicyclo-2-2-1-heptane-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P45510
Availability details for
Quantity
  • Availability
  • Add to Cart