ADVH1F1C5C64 AChemBlock

(2S,3S)-1-benzhydryl-2-methyl-azetidin-3-amine

  • MDL number MFCD31705181
  • Cas Number : 164906-71-2
  • Molecular Weight : 252.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2s-3s-1-benzhydryl-2-methyl-azetidin-3-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P47415
Availability details for
Quantity
  • Availability
  • Add to Cart