ADVH1F1C5C86 AChemBlock

8-methyl-1,4-dioxaspiro[4.5]decane-8-carboxylic acid

  • MDL number MFCD18374792
  • Cas Number : 412293-42-6
  • Molecular Weight : 200.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/8-methyl-1-4-dioxaspiro-4-5-decane-8-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P45638
Availability details for
Quantity
  • Availability
  • Add to Cart