ADVH1F1C5C8A AChemBlock

2-(1,4-dioxa-spiro[4.5]dec-8-yl)-ethylamine

  • MDL number MFCD04114534
  • Cas Number : 124499-34-9
  • Molecular Weight : 185.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-1-4-dioxa-spiro-4-5-dec-8-yl-ethylamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P49197
Availability details for
Quantity
  • Availability
  • Add to Cart