ADVH1F1C5CB3 AChemBlock

(1r,4r)-4-({[(tert-butoxy)carbonyl]amino}methyl)cyclohexane-1-carboxylic acid

  • MDL number MFCD00673773
  • Cas Number : 27687-14-5
  • Molecular Weight : 257.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-4r-4-tert-butoxy-carbonyl-amino-methyl-cyclohexane-1-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P45843
Availability details for
Quantity
  • Availability
  • Add to Cart