ADVH1F1C5CF0 AChemBlock

5-Fluoro-1H-pyrazolo[3,4-b]pyridine-3-carboxylic acid

  • MDL number MFCD18260736
  • Cas Number : 1211586-74-1
  • Molecular Weight : 181.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-fluoro-1h-pyrazolo-3-4-b-pyridine-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P44119
Availability details for
Quantity
  • Availability
  • Add to Cart