ADVH1F1C5D8E AChemBlock

tert-butyl (1R,4S,6R)-6-hydroxy-2-azabicyclo[2.2.1]heptane-2-carboxylate

  • MDL number MFCD22209823
  • Cas Number : 1932398-86-1
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1r-4s-6r-6-hydroxy-2-azabicyclo-2-2-1-heptane-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P46639
Availability details for
Quantity
  • Availability
  • Add to Cart