ADVH1F1C5E09 AChemBlock

6-fluoro-1H-pyrrolo[3,2-b]pyridine-3-carboxylic acid

  • MDL number MFCD12962612
  • Cas Number : 1190316-18-7
  • Molecular Weight : 180.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-fluoro-1h-pyrrolo-3-2-b-pyridine-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P49551
Availability details for
Quantity
  • Availability
  • Add to Cart