ADVH1F1C5E43 AChemBlock

2-(azetidin-1-yl)propan-1-ol

  • MDL number MFCD30802779
  • Cas Number : 1849348-02-2
  • Molecular Weight : 115.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-azetidin-1-yl-propan-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P46846
Availability details for
Quantity
  • Availability
  • Add to Cart