ADVH1F1C5EBF AChemBlock

5-(methoxycarbonyl)bicyclo[3.1.1]heptane-1-carboxylic acid

  • MDL number MFCD00220676
  • Cas Number : 110371-28-3
  • Molecular Weight : 198.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-methoxycarbonyl-bicyclo-3-1-1-heptane-1-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P49802
Availability details for
Quantity
  • Availability
  • Add to Cart