ADVH1F1C604C AChemBlock

N-ethylcyclobutanamine

  • MDL number MFCD16107849
  • Cas Number : 852108-24-8
  • Molecular Weight : 99.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/n-ethylcyclobutanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P45089
Availability details for
Quantity
  • Availability
  • Add to Cart