ADVH1F1C6106 AChemBlock

(1S,2S)-2-(methoxymethyl)cyclobutan-1-amine hydrochloride

  • MDL number MFCD31705646
  • Cas Number : 1807937-89-8
  • Molecular Weight : 151.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1s-2s-2-methoxymethyl-cyclobutan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P46994
Availability details for
Quantity
  • Availability
  • Add to Cart