ADVH1F1C617E AChemBlock

tert-butyl (1R,5S)-3,6-diazabicyclo[3.1.1]heptane-6-carboxylate

  • MDL number MFCD17016736
  • Cas Number : 869494-16-6
  • Molecular Weight : 198.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1r-5s-3-6-diazabicyclo-3-1-1-heptane-6-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P50137
Availability details for
Quantity
  • Availability
  • Add to Cart