ADVH1F1C623D AChemBlock

8-oxobicyclo[3.2.1]octane-3-carboxylic acid

  • MDL number MFCD09837131
  • Cas Number : 340164-19-4
  • Molecular Weight : 168.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/8-oxobicyclo-3-2-1-octane-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P45735
Availability details for
Quantity
  • Availability
  • Add to Cart