ADVH1F1C6407 AChemBlock

3-(difluoromethyl)bicyclo[1.1.1]pentane-1-carboxylic acid

  • MDL number MFCD29767889
  • Cas Number : 2090481-18-6
  • Molecular Weight : 162.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp.
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-difluoromethyl-bicyclo-1-1-1-pentane-1-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P46355
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart