ADVH1F1C643C AChemBlock

ethyl 1,4-diazabicyclo[2.2.2]octane-2-carboxylate

  • MDL number MFCD20492154
  • Cas Number : 29924-70-7
  • Molecular Weight : 184.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-1-4-diazabicyclo-2-2-2-octane-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P45795
Availability details for
Quantity
  • Availability
  • Add to Cart