ADVH1F1C6493 AChemBlock

1-azabicyclo[2.2.2]octane-2-carboxylic acid hydrochloride

  • MDL number MFCD00168044
  • Cas Number : 52601-23-7
  • Molecular Weight : 191.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-azabicyclo-2-2-2-octane-2-carboxylic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P45512
Availability details for
Quantity
  • Availability
  • Add to Cart