ADVH1F1C64FF AChemBlock

{3-bromobicyclo[1.1.1]pentan-1-yl}methanol

  • MDL number MFCD31705883
  • Cas Number : 137741-16-3
  • Molecular Weight : 177.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-bromobicyclo-1-1-1-pentan-1-yl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P48511
Availability details for
Quantity
  • Availability
  • Add to Cart