ADVH1F1C6562 AChemBlock

2-methyl-1H-pyrrolo[2,3-c]pyridine-3-carboxylic acid

  • MDL number MFCD18810477
  • Cas Number : 933736-80-2
  • Molecular Weight : 176.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-1h-pyrrolo-2-3-c-pyridine-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P44810
Availability details for
Quantity
  • Availability
  • Add to Cart