ADVH1F1C656D AChemBlock

2-(azetidin-3-ylidene)acetonitrile hydrochloride

  • MDL number MFCD19227479
  • Cas Number : 1314910-43-4
  • Molecular Weight : 130.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-azetidin-3-ylidene-acetonitrile-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P44064
Availability details for
Quantity
  • Availability
  • Add to Cart