ADVH303B9544 AChemBlock

tert-butyl N-[endo-3-azabicyclo[3.1.0]hexan-6-yl]carbamate

  • Cas Number : 134677-60-4
  • Molecular Weight : 198.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-endo-3-azabicyclo-3-1-0-hexan-6-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q51861
Availability details for
Quantity
  • Availability
  • Add to Cart