ADVH303B9554 AChemBlock

4-chloro-7-methyl-7H-pyrrolo[2,3-d]pyrimidine-5-carboxylic acid

  • MDL number MFCD09909362
  • Cas Number : 1069473-61-5
  • Molecular Weight : 211.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-7-methyl-7h-pyrrolo-2-3-d-pyrimidine-5-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q51880
Availability details for
Quantity
  • Availability
  • Add to Cart