ADVH303B9580 AChemBlock

6-bromo-2-methyl-2H-indazol-5-ol

  • MDL number MFCD23106323
  • Cas Number : 1403767-25-8
  • Molecular Weight : 227.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-2-methyl-2h-indazol-5-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q51940
Availability details for
Quantity
  • Availability
  • Add to Cart