ADVH303B9585 AChemBlock

3-((benzyloxy)carbonyl)-3-azabicyclo[3.1.0]hexane-6-carboxylic acid

  • Cas Number : 134677-59-1
  • Molecular Weight : 261.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-benzyloxy-carbonyl-3-azabicyclo-3-1-0-hexane-6-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q51945
Availability details for
Quantity
  • Availability
  • Add to Cart