ADVH303B962E AChemBlock

3,3-difluoro-4-methoxypyrrolidine hydrochloride

  • MDL number MFCD27987047
  • Cas Number : 1638764-85-8
  • Molecular Weight : 173.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-3-difluoro-4-methoxypyrrolidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q52144
Availability details for
Quantity
  • Availability
  • Add to Cart