ADVH303B9668 AChemBlock

1-(fluoromethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

  • MDL number MFCD28401077
  • Cas Number : 1446481-36-2
  • Molecular Weight : 226.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-fluoromethyl-4-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-1h-pyrazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q52206
Availability details for
Quantity
  • Availability
  • Add to Cart