ADVH303B96B9 AChemBlock

4-(bromomethyl)-2,2,6,6-tetramethyltetrahydro-2H-pyran

  • MDL number MFCD28501353
  • Cas Number : 1638760-06-1
  • Molecular Weight : 235.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-bromomethyl-2-2-6-6-tetramethyltetrahydro-2h-pyran.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q52296
Availability details for
Quantity
  • Availability
  • Add to Cart