ADVH303B96C0 AChemBlock

tert-butyl 3-(aminomethyl)-5-fluoropiperidine-1-carboxylate

  • MDL number MFCD22690367
  • Cas Number : 1374655-64-7
  • Molecular Weight : 232.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-aminomethyl-5-fluoropiperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q52303
Availability details for
Quantity
  • Availability
  • Add to Cart