ADVH303B96EF AChemBlock

(R)-1-(tert-butoxycarbonyl)-3-methylpiperidine-3-carboxylic acid

  • MDL number MFCD28501395
  • Cas Number : 1415018-76-6
  • Molecular Weight : 243.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/r-1-tert-butoxycarbonyl-3-methylpiperidine-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q52352
Availability details for
Quantity
  • Availability
  • Add to Cart