ADVH303B96FA AChemBlock

(3R,5S)-5-methylpiperidine-3-carboxylic acid

  • MDL number MFCD28501405
  • Cas Number : 1638743-96-0
  • Molecular Weight : 143.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3r-5s-5-methylpiperidine-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q52364
Availability details for
Quantity
  • Availability
  • Add to Cart