ADVH303B978C AChemBlock

methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinate

  • MDL number MFCD08752647
  • Cas Number : 957062-72-5
  • Molecular Weight : 263.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-picolinate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q52524
Availability details for
Quantity
  • Availability
  • Add to Cart