ADVH303B97E2 AChemBlock

(3R,4S)-4-methoxypyrrolidin-3-ol hydrochloride

  • MDL number MFCD28501501
  • Cas Number : 473298-26-9
  • Molecular Weight : 153.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3r-4s-4-methoxypyrrolidin-3-ol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q52626
Availability details for
Quantity
  • Availability
  • Add to Cart