ADVH303B981C AChemBlock

cis-tert-butyl 3-amino-2-methylpiperidine-1-carboxylate

  • Cas Number : 1628258-95-6
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/cis-tert-butyl-3-amino-2-methylpiperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q52686
Availability details for
Quantity
  • Availability
  • Add to Cart