ADVH303B989E AChemBlock

(R)-4,4-difluoropyrrolidin-3-ol hydrochloride

  • Cas Number : 1433363-34-8
  • Molecular Weight : 159.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/r-4-4-difluoropyrrolidin-3-ol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q52826
Availability details for
Quantity
  • Availability
  • Add to Cart