ADVH303B9ACE AChemBlock

4-bromo-3-chloro-2-methylpyridine

  • MDL number MFCD11215565
  • Cas Number : 1188140-52-4
  • Molecular Weight : 206.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-3-chloro-2-methylpyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q53251
Availability details for
Quantity
  • Availability
  • Add to Cart