ADVH303B9AE2 AChemBlock

3-methyleneoxetane

  • MDL number MFCD31728327
  • Cas Number : 10242-62-3
  • Molecular Weight : 70.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-methyleneoxetane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q53034
Availability details for
Quantity
  • Availability
  • Add to Cart