ADVH303B9B37 AChemBlock

2-bromo-5-fluoro-4-nitropyridine

  • MDL number MFCD28740491
  • Cas Number : 1805578-52-2
  • Molecular Weight : 220.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-bromo-5-fluoro-4-nitropyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q53373
Availability details for
Quantity
  • Availability
  • Add to Cart