ADVH303B9B3B AChemBlock

1-benzyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

  • MDL number MFCD31729402
  • Cas Number : 1430753-39-1
  • Molecular Weight : 334.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-benzyl-5-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-1h-indazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q53377
Availability details for
Quantity
  • Availability
  • Add to Cart