ADVH303B9BAD AChemBlock

3-bromo-6-(trifluoromethoxy)pyridin-2-amine

  • MDL number MFCD25450895
  • Cas Number : 1361852-29-0
  • Molecular Weight : 257.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-6-trifluoromethoxy-pyridin-2-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q53518
Availability details for
Quantity
  • Availability
  • Add to Cart