ADVH303B9BE2 AChemBlock

rel-(2R,3S)-piperidine-2,3-dicarboxylic acid

  • Cas Number : 46026-75-9
  • Molecular Weight : 173.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/rel-2r-3s-piperidine-2-3-dicarboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q53601
Availability details for
Quantity
  • Availability
  • Add to Cart