ADVH303B9C9D AChemBlock

2-amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinonitrile

  • MDL number MFCD16995200
  • Cas Number : 1246372-66-6
  • Molecular Weight : 245.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-amino-5-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-nicotinonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q53831
Availability details for
Quantity
  • Availability
  • Add to Cart