ADVH303B9DE5 AChemBlock

1-(4-methoxybenzyl)-5-methyl-4-nitro-1H-pyrazol-3-amine

  • MDL number MFCD31742959
  • Cas Number : 2227204-81-9
  • Molecular Weight : 262.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-4-methoxybenzyl-5-methyl-4-nitro-1h-pyrazol-3-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q54424
Availability details for
Quantity
  • Availability
  • Add to Cart