ADVH303B9EC4 AChemBlock

tert-butyl (2R,4R)-2-cyano-4-hydroxypyrrolidine-1-carboxylate

  • Cas Number : 1932029-59-8
  • Molecular Weight : 212.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2r-4r-2-cyano-4-hydroxypyrrolidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q54352
Availability details for
Quantity
  • Availability
  • Add to Cart