ADVH303B9F7F AChemBlock

7-bromo-5-fluoro-1-methyl-1H-indazole

  • MDL number MFCD29089688
  • Cas Number : 1781864-19-4
  • Molecular Weight : 229.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-5-fluoro-1-methyl-1h-indazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q54554
Availability details for
Quantity
  • Availability
  • Add to Cart