ADVH303B9FFE AChemBlock

tert-butyl 6-benzyl-2,6-diazaspiro[3.4]octane-2-carboxylate

  • MDL number MFCD21099567
  • Cas Number : 1352926-14-7
  • Molecular Weight : 302.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-6-benzyl-2-6-diazaspiro-3-4-octane-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q54693
Availability details for
Quantity
  • Availability
  • Add to Cart