ADVH303BA045 AChemBlock

6-(tert-butoxycarbonyl)-2-oxa-6-azaspiro[3.4]octane-8-carboxylic acid

  • MDL number MFCD28954136
  • Cas Number : 1803608-20-9
  • Molecular Weight : 257.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-tert-butoxycarbonyl-2-oxa-6-azaspiro-3-4-octane-8-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q54769
Availability details for
Quantity
  • Availability
  • Add to Cart