ADVH303BA125 AChemBlock

tert-butyl 9-hydroxy-3-azaspiro[5.5]undecane-3-carboxylate

  • MDL number MFCD12755293
  • Cas Number : 918644-73-2
  • Molecular Weight : 269.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-9-hydroxy-3-azaspiro-5-5-undecane-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q55003
Availability details for
Quantity
  • Availability
  • Add to Cart