ADVH303BA15D AChemBlock

(R)-3,3-difluorocyclopentan-1-ol

  • Cas Number : 2090758-49-7
  • Molecular Weight : 122.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/r-3-3-difluorocyclopentan-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q55062
Availability details for
Quantity
  • Availability
  • Add to Cart