ADVH303BA1AB AChemBlock

5-methoxy-1,3-dihydro-2H-pyrrolo[2,3-b]pyridin-2-one

  • MDL number MFCD12962880
  • Cas Number : 1190322-28-1
  • Molecular Weight : 164.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-methoxy-1-3-dihydro-2h-pyrrolo-2-3-b-pyridin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q55141
Availability details for
Quantity
  • Availability
  • Add to Cart